Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNJ2BP/ARIP2 Antibody (OTI3A9), FITC, Novus Biologicals™
SDP

Catalog No. NBP274417F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274417F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274417F Supplier Novus Biologicals Supplier No. NBP274417F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYNJ2BP/ARIP2 Monoclonal specifically detects SYNJ2BP/ARIP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen SYNJ2BP/ARIP2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3A9
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias activin receptor interacting protein 5, ARIP2, FLJ11271, Mitochondrial outer membrane protein 25, OMP25FLJ41973, synaptojanin 2 binding protein, synaptojanin-2-binding protein
Gene Symbols SYNJ2BP
Host Species Mouse
Immunogen Full length recombinant protein of human SYNJ2BP/ARIP2 (NP_060843) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55333.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.