Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synphilin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $437.00
Specifications
Antigen | Synphilin 1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145488
|
Invitrogen™
PA5145488 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145487
|
Invitrogen™
PA5145487 |
200 μL |
Each for $437.00
|
|
|||||
Description
Parkinson is the second most common neurodegenerative disease after Alzheimer's. About 1 percent of people over the age of 65 and 3 percent of people over the age of 75 are affected by the disease. The mutation is the most common cause of Parkinson disease identified to date. Synuclein alpha interacting protein (Synphilin-1) contains several protein-protein interaction domains and interacts with alpha synuclein in neurons. Mutations of SNCAIP have been linked to Parkinson disease. The amino acid sequence of synphilin-1 shows extensive homology with its human counterpart, especially in regions containing ankyrin-like motifs and the coiled-coil domain. Expression of mouse synphilin-1 in tissues is similar to its human counterpart. Synphilin-1 has an important role in the formation of aggregates and cytotoxicity in Parkinson disease and that Dorfin may be involved in the pathogenic process by ubiquitylation of synphilin-1. Role of synphilin-1 in synaptic function and protein degradation and in the molecular mechanisms leading to neurodegeneration in Parkinson disease.Specifications
Synphilin 1 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
Q99ME3 | |
307309, 67847 | |
Recombinant Rat SNCaIP1 protein, Pro766-Leu963 (Accession #D3ZWQ5). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
2810407O15Rik; 4933427B05Rik; Alpha-synuclein-interacting protein; BB104594; MGC39814; SNCAIP; Sph1; Synphilin 1; synphilin-1; synuclein alpha interacting protein; synuclein, alpha interacting protein; synuclein, alpha interacting protein (synphilin); SYPH1 | |
SNCAIP | |
IgG | |
Affinity chromatography | |
SNCAIP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title