Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNPO2L Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017332 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017332 Supplier Novus Biologicals Supplier No. NBP282021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYNPO2L Polyclonal specifically detects SYNPO2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SYNPO2L
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias Synaptopodin 2-Like, Synaptopodin 2-Like Protein
Gene Symbols SYNPO2L
Host Species Rabbit
Immunogen SYNPO2L antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SYNPO2L. The immunogen is located within amino acids 140 - 190 of SYNPO2L. Amino Acid Squence: PATQEKPRRPRRRGPTRPT
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79933
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.