Learn More
Syntaxin 1 Polyclonal Antibody, Invitrogen™
Description
PA1-1042 detects Syntaxin 1 in rat, mouse and bovine samples. PA1-1042 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ∽33 kDa protein representing Syntaxin 1 in rat, mouse and bovine brain tissue samples. The PA1-1042 immunogen is a synthetic peptide corresponding to residues V(22) T V D R D R F M D E F F E Q V E E I R(41) of rat Syntaxin 1. This sequence is conserved in mouse, human and bovine. The PA1-1042 immunizing peptide (Cat. No. PEP-278) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Syntaxin 1 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | STX1A |
| Gene Accession No. | P32850, Q16623, O35526, P32851 |
| Gene Alias | HPC-1, P35-1, STX1, SYN1A |
| Gene Symbols | STX1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.