Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 10 Mouse anti-Human, Clone: 1B1C3, Proteintech
SDP

Catalog No. 501737211 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7211 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7211 Supplier Proteintech Group Inc Supplier No. 668211IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNARE involved in vesicular transport from the late endosomes to the trans-Golgi network.

Specifications

Antigen Syntaxin 10
Applications Western Blot
Classification Monoclonal
Clone 1B1C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene STX10
Gene Accession No. O60499
Gene Alias hsyn10, STX10, SYN10, syntaxin 10
Gene Symbols STX10
Host Species Mouse
Immunogen Syntaxin 10 Fusion Protein Ag27861
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8677
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.