Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin-BP1, p Ser513 Antibody, Novus Biologicals™
SDP

Catalog No. NB300742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300742 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300742 Supplier Novus Biologicals Supplier No. NB300742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Syntaxin-BP1 Polyclonal specifically detects Syntaxin-BP1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Syntaxin-BP1 (p Ser513)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q62753
Gene Alias EIEE4, FLJ37475, hUNC18, MUNC18-1, N-Sec1, NSEC1, P67, Protein unc-18 homolog 1, Protein unc-18 homolog A, RBSEC1, syntaxin binding protein 1, syntaxin-binding protein 1, Unc18-1, unc-18A, UNC18A, UNC18neuronal SEC1
Gene Symbols STXBP1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser513 conjugated to KLH. Accession # Q62753
Molecular Weight of Antigen 65 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6812
Test Specificity Specific for the ∼65k Munc-18 protein phosphorylated at Ser515. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.