Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntenin 1 Antibody (K10P3D5*H3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250287UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250287UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250287UV Supplier Novus Biologicals Supplier No. NBP250287UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Syntenin 1 Monoclonal specifically detects Syntenin 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Syntenin 1
Applications Western Blot
Classification Monoclonal
Clone K10P3D5*H3
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1
Gene Symbols SDCBP
Host Species Mouse
Immunogen Peptide - HIIKRMAPS
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6386
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.