Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntenin 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300835 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300835 Supplier Novus Biologicals Supplier No. NB300835
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Syntenin 2 Polyclonal specifically detects Syntenin 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Syntenin 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 4 - 6 ug/ml, Immunohistochemistry-Paraffin 4 - 6 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_056500
Gene Alias FLJ12256, SITAC18ST-2, syndecan binding protein (syntenin) 2, Syndecan-binding protein 2, syntenin-2
Gene Symbols SDCBP2
Host Species Goat
Immunogen Peptide with sequence C-LLHHTMDHSIPDA corresponding to C-Terminus according to NP_056500.3, NP_536737.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27111
Test Specificity This antibody is expected to recognize both human isoforms (NP_056500; NP_536737).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.