Learn More
Synthware™ Distillation Receiver, Cow Type, with Hose Connection
With hose connection for application of vacuum
$169.20 - $192.00
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
31-501-030
![]() ![]() |
Synthware™
D681420 |
14/20 |
Each for $178.80
|
|
|||||
31-501-031
![]() ![]() |
Synthware™
D681922 |
19/22 |
Each for $169.20
|
|
|||||
31-501-032
![]() ![]() |
Synthware™
D682440 |
24/40 |
Each for $192.00
|
|
|||||
Description
- With three receivers spaced 40 deg. apart at 105 deg. angle between top outer joint and lower inner drip tip joints
- All joints of same size
Specifications
Cow Type Distillation Receiver |