Learn More
Synthware™ Distilling Trap Adapters
GREENER_CHOICESDP
Placed above flask to prevent carry-over of reactions which have a tendency to foam
$92.28 - $99.96
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Product Type Distilling Trap Adapter
For Use With (Application) Organic Synthesis
Quantity 1
View More Specs
Products 3
Catalog Number Mfr. No. Standard Taper Joint Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Price Quantity & Availability  
31-500-609
GREENER_CHOICESDP
View Documents
Synthware™
A262414
Upper: 24/40, Lower: 14/20
Each for $92.28
Only null left
Add to Cart
31-500-610
GREENER_CHOICESDP
View Documents
Synthware™
A262419
Upper: 24/40, Lower: 19/22
Each for $95.04
Only null left
Add to Cart
31-500-611
GREENER_CHOICESDP
View Documents
Synthware™
A264100
Upper: 24/40, Lower: 24/40
Each for $99.96
Only null left
Add to Cart
Description
Description
Synthware Kjeldahl distilling trap adapter. Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top outer and lower inner standard taper joints.
Specifications
Specifications
100 mL
Distilling Trap Adapter
1
Borosilicate Glass (3.3)
Organic Synthesis
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.