Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Low Form Gooch Crucibles
GREENER_CHOICESDP

Catalog No. 31501444 Shop All Kemtech Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
Porosity:
Coarse
Medium

Crucibles include a fritted disc in varying porosities.

  • Low form gooch crucible with fritted disc
  • Available in medium or coarse porosities

Specifications

Color Clear
Description Gooch Crucible
Material Glass
For Use With (Application) Organic Synthesis
Quantity 1
Capacity (Metric) 30 mL
Diameter (Metric) Outer 30 mm
Product Type Gooch crucible
Porosity Medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.