Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntrophin alpha 1 Antibody (OTI1H10), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274422A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274422A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274422A70 Supplier Novus Biologicals Supplier No. NBP274422AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Syntrophin alpha 1 Monoclonal specifically detects Syntrophin alpha 1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Syntrophin alpha 1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H10
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin
Gene Symbols SNTA1
Host Species Mouse
Immunogen Full length recombinant protein of human Syntrophin alpha 1 (NP_003089) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6640
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.