Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein alpha/beta Antibody (3B6), FITC, Novus Biologicals™
SDP

Catalog No. NBP122957F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122957F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122957F Supplier Novus Biologicals Supplier No. NBP122957F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synuclein alpha/beta Monoclonal specifically detects Synuclein alpha/beta in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Synuclein alpha/beta
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3B6
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta-synuclein, I(2)-synuclein, I+/--synuclein, NACP, non A-beta component of AD amyloid, PARK1, PD1, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Gene Symbols SNCA
Host Species Mouse
Immunogen This Synuclein alpha/beta Antibody (3B6) was developed against recombinant human Synuclein alpha/beta (119-140aa) purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6622
Test Specificity Anti-human alpha-synuclein (alpha-synuclein 119-140aa) mAb, clone 3B6, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human alpha-synuclein protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.