Learn More
MilliporeSigma™ Supelco™ Syringe for On-Column Injection
Catalog No. p-7213569
Click to view available options
For Use With (Equipment):
Agilent/HP Injectors
In this system, a 32-gauge needle is inserted in a cool, narrow bore injection valve and then into capillary column
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 32 G
Diameter (Metric) Outer Needle 0.24 mm
For Use With (Application) On-Column Injection
Product Type Syringe
Needle Point Number 3
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 12.5 cm
Needle Point Style Blunt
For Use With (Equipment) Agilent/HP Injectors
Connection Type Fixed
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.