Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT7 Antibody (S275-14), FITC, Novus Biologicals™
SDP

Catalog No. NBP222420F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222420F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222420F Supplier Novus Biologicals Supplier No. NBP222420F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYT7 Monoclonal specifically detects SYT7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SYT7
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S275-14
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias IPCA-7synaptotagmin 7, MGC150517, prostate cancer associated protein 7, Prostate cancer-associated protein 7, synaptotagmin VIIPCANAP7, synaptotagmin-7, sytVII, SYT-VII
Gene Symbols SYT7
Host Species Mouse
Immunogen Fusion protein amino acids 150-239 (Cytoplasmic C2A domain) of mouse Synaptotagmin-7
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9066
Test Specificity Detects approx 45kDa. Does not cross-react with Synaptotagmin-6 (or others). Can identify other isoforms bands at approx 65kD.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.