Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SZRD1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB326689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326689 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326689 Supplier Novus Biologicals Supplier No. NBP299181APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SZRD1 Polyclonal antibody specifically detects SZRD1 in Human samples. It is validated for Western Blot

Specifications

Antigen SZRD1
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias C1orf144, chromosome 1 open reading frame 144, DKFZp566C0424, MGC70432, PM21, putative MAPK activating protein PM20, Putative MAPK-activating protein PM18/PM20/PM22
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SZRD1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26099
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.