Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ T 0070907

Click to view available options
Quantity:
5 mg
25 mg
Description
T 0070907 is a potent and selective irreversible PPARγ antagonist (IC50 = 1 nM). Displays > 800-fold selectivity for PPARγ over PPARα and PPARδ. Blocks transcriptional activity of PPARγ in vitro and inhibits Rosiglitazone -induced adipogenesis.
Specifications
Specifications
Chemical Name or Material | 2-Chloro-5-nitro-N-4-pyridinylbenzamide |
CAS | 313516-66-4 |
Assay Percent Range | >99% |
Quantity | 25 mg |
Molecular Formula | C12H8N3O3Cl |
Formula Weight | Observed MW: 277.67 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction