Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ T 0901317
GREENER_CHOICE

Catalog No. p-4784627
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent liver X receptor (LXR) agonist; also CAR inverse agonist

Potent, high affinity liver X receptor (LXR) agonist (EC50 ∽ 50 nM, Kd values are 7 and 22 nM for LXR-α and LXR-β respectively). Upregulates expression of the ABCA1 gene associated with cholesterol efflux regulation and HDL metabolism. Decreases amyloid-β production in primary neurons in vitro. Displays an EC50 of ∽ 5 μM for activation of bile acid farnesoid X receptors (FXRs); 10-fold more potent than natural FXR ligand chenodeoxycholic acid. Also exhibits inverse agonist activity at constitutive androstane receptors (CAR).

Chemical Identifiers

CAS 293754-55-9
Molecular Formula C17H12F9NO3S

Specifications

Quantity 10 mg
Formula Weight 481.33
Purity >98%
Molecular Weight (g/mol) 481.333
Chemical Name or Material T 0901317
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.