Learn More
T- and B-Cell Activation Antigen Rat anti-Mouse, FITC, Clone: GL7, BD
Rat Monoclonal Antibody
$116.00 - $404.00
Specifications
Antigen | T- and B-Cell Activation Antigen |
---|---|
Clone | GL7 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB562080
|
BD Biosciences
562080 |
50 μg |
Each for $116.00
|
|
|||||
BDB553666
|
BD Biosciences
553666 |
0.5 mg |
Each for $404.00
|
|
|||||
Description
The GL7 antibody reacts with a 35-kDa cell-surface protein found on T and B lymphocytes activated in vitro,on bone marrow Pre-B-II cells, germinal-center B cells, and a subpopulation of the thymocyte fraction expressing high density of CD3e. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6.
Specifications
T- and B-Cell Activation Antigen | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
GL7 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgM κ | |
Affinity Purified | |
GL7 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.