Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet, Mouse Anti-House RY703, Clone: O4-46, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 571477
Description
T-bet Monoclonal antibody specifically detects T-bet in Human (QC Testing), Mouse (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
T-bet | |
Monoclonal | |
RY703 | |
T-box expressed in T cells; TBX21; T-box 21; TBLYM | |
Human T-bet Peptide | |
100 Tests | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry (Intracellular Staining) | |
O4-46 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
30009, 57765 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction