Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet, Mouse Anti-House RY703, Clone: O4-46, BD Horizon™
Mouse Monoclonal Antibody
$216.00 - $442.00
Specifications
Antigen | T-bet |
---|---|
Clone | O4-46 |
Applications | Flow Cytometry (Intracellular Staining) |
Classification | Monoclonal |
Conjugate | RY703 |
Description
T-bet Monoclonal antibody specifically detects T-bet in Human (QC Testing), Mouse (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
T-bet | |
Flow Cytometry (Intracellular Staining) | |
RY703 | |
Mouse | |
Human, Mouse | |
T-box expressed in T cells; TBX21; T-box 21; TBLYM | |
Human T-bet Peptide | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
O4-46 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
30009, 57765 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title