Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-bet/TBX21 Antibody (4B10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP143298V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143298V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143298V Supplier Novus Biologicals Supplier No. NBP143298V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).

Specifications

Antigen T-bet/TBX21
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 4B10
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Flow (Intracellular)
Gene Alias T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Gene Symbols TBX21
Host Species Mouse
Immunogen The immunogen for this antibody was TBet.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30009
Target Species Human, Mouse, Rhesus Macaque
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.