Learn More
Description
Specifications
Specifications
| Antigen | T-bet/TBX21 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4B10 |
| Conjugate | DyLight 650 |
| Dilution | Western Blot, Flow Cytometry, Flow (Intracellular) |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM |
| Gene Symbols | TBX21 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
