Learn More
Description
Specifications
Specifications
| Antigen | T-bet/TBX21 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4B10 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Flow Cytometry, Flow (Intracellular) |
| Gene Alias | T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM |
| Gene Symbols | TBX21 |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody was TBet. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
