Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-bet/TBX21 Antibody (525831), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347358 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347358 Supplier Novus Biologicals Supplier No. FAB53851MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen T-bet/TBX21
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 525831
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Host Species Mouse
Immunogen E. coli-derived recombinant human T-bet/TBX21, Glu326-Asn535, Accession # Q9UL17
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 30009
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.