Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-box 19 Antibody (CL6251), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | T-box 19 |
---|---|
Clone | CL6251 |
Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
T-box 19 Monoclonal specifically detects T-box 19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
T-box 19 | |
Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Unconjugated | |
Mouse | |
dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543 | |
TBX19 | |
IgG1 | |
Protein A purified |
CL6251 | |
Monoclonal | |
Purified | |
Human | |
9095 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EVHASTPGAFLLGNPAVTSPPSVLSTQAPTSAGVEVLGEPSLTSIAVSTWTAVASHPFAGWGGPGAGGHHSPSSLDG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title