Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-box 19 Antibody (OTI1C9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274469R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274469R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274469R Supplier Novus Biologicals Supplier No. NBP274469R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

T-box 19 Monoclonal specifically detects T-box 19 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen T-box 19
Applications Western Blot
Classification Monoclonal
Clone OTI1C9
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543
Gene Symbols TBX19
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-238 of human T-box 19 (NP_005140) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9095.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.