Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

t-Plasminogen Activator/tPA Antibody (OTI3H3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274568A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274568A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274568A647 Supplier Novus Biologicals Supplier No. NBP274568AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

t-Plasminogen Activator/tPA Monoclonal specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen t-Plasminogen Activator/tPA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3H3
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, t-PA, TPA, t-plasminogen activator
Gene Symbols PLAT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 157-426 of human t-Plasminogen Activator/tPA (NP_000921) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5327.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.