Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences T4 DNA Polymerase
SCI_ED

T4 DNA Polymerase is used for 3'-overhang removal to form blunt ends, 5'-overhang fill-in to form blunt ends, probe labeling and DNA library preparation for Next-generation sequencing.

$60.00 - $375.00

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance Low
PCR Method Standard PCR, End-point PCR, qPCR, Multiplex qPCR
Reaction Speed Standard
View More Specs

Products 3
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
S03108
SCI_ED
View Documents
G-Biosciences
7862006
1500
Each for $375.00
Only null left
Add to Cart
 
S03110
SCI_ED
View Documents
G-Biosciences
7862005
750
Each for $214.00
Only null left
Add to Cart
 
S03112
SCI_ED
View Documents
G-Biosciences
7862004
200
Each for $60.00
Only null left
Add to Cart
 
Description

Description

  • T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' - 3' direction with the presence of template and primer.
  • It has both DNA-dependent DNA polymerase activity and 3' - 5' exonuclease activity which make the enzyme useful for several molecular biology applications.
Specifications

Specifications

Gel Electrophoresis
Low
Standard
DNA Polymerase
Liquid
Standard PCR, End-point PCR, qPCR, Multiplex qPCR
Biotin and Other Hapten, No ROX, Biotin, Unlabeled, Unconjugated
Pathogen Detection, Non-Radioactive Labeling, DNA Sequencing, Next-Generation Sequencing (NGS), Cloning
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.