Learn More
T4 UvsX DNA Recombinase, Lyophilized, G-Biosciences

T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.
$355.00 - $1850.00
Specifications
Detection Method | Gel Electrophoresis |
---|---|
Form | Lyophilized |
GC-Rich PCR Performance | High |
PCR Method | End-point PCR |
Reaction Speed | 20 min. |
Description
The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesized strand displacing the old strand. The newly synthesized duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA
Specifications
Gel Electrophoresis | |
High | |
20 min. | |
Microbial Detection, Pathogen Detection, Isothermal Amplification |
Lyophilized | |
End-point PCR | |
Biotin, FAM, TAMRA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.