Learn More
T4 UvsX DNA Recombinase, Lyophilized, G-Biosciences
T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.
Supplier: G-Biosciences 7862136
Description
The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesized strand displacing the old strand. The newly synthesized duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA
Specifications
Gel Electrophoresis | |
High | |
20 min. | |
Uvsx recombinase | |
Microbial Detection, Pathogen Detection, Isothermal Amplification |
Lyophilized | |
End-point PCR | |
Biotin, FAM, TAMRA | |
1000 μg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.