Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 Epitope Tag Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB600377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600377 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600377 Supplier Novus Biologicals Supplier No. NB600377
Add to Cart
Add to Cart

Goat Polyclonal Antibody

T7 Epitope Tag Polyclonal antibody specifically detects T7 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.

Specifications

Antigen T7 Epitope Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000-1:10000, ELISA 1:10000-1:100000, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunohistochemistry (Negative)
Formulation PBS and 0.2% BSA with 0.1% Pro-Clean 400
Gene Alias MASMTGGQQMG epitope tag, MASMTGGQQMG tag, T7 epitope tag
Host Species Goat
Immunogen Goats were immunized with T7 cleavage site (MASMTGGQQMG) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, T7 Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.