Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAB-2 Mouse anti-Human, Clone: 1D5.2, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | TAB-2 |
---|---|
Clone | 1D5.2 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS1894-25
|
MilliporeSigma
MABS189425UG |
25 μg |
Each for $192.35
|
|
|||||
MABS1894-100
|
MilliporeSigma
MABS1894100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-TAB-2, clone 1D5.2 is a mouse monoclonal antibody that detects TAK1-binding protein 2 and has been tested for use in Western Blotting.Specifications
TAB-2 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
TAB2,KIAA0733,MAP3K7IP2 | |
IgG2b κ | |
Protein G Purified | |
Clone 1D5.2 specifically detects human TGF-beta-activated kinase 1 and MAP3K7-binding protein 2. It targets an epitope within 133 amino acids from the C-terminal region. |
1D5.2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Signaling | |
NP_001278963 | |
GST/His-tagged recombinant fragment corresponding to 133 amino acids from the C-terminal region of human TAK1-binding protein 2 (TAB-2) | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title