Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAB1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB198094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198094 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198094 Supplier Novus Biologicals Supplier No. NBP176595R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAB1 Polyclonal antibody specifically detects TAB1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TAB1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias MAP3K7IP13'-Tab1, MGC57664, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TGF-beta-activated kinase 1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 13 amino acids in the center of human TAB1. The immunogen is located within amino acids 220 - 270 of TAB1. Amino Acid Squence: ESTRRIGDYKVKY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10454
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.