Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAB1 Rabbit anti-Human, Mouse, Rat, Clone: 2W0P2, Novus Biologicals™
SDP

Catalog No. NB167426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167426 20 μg
NB167425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167426 Supplier Novus Biologicals Supplier No. NBP31616720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TAB1 Monoclonal antibody specifically detects TAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen TAB1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 2W0P2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias MAP3K7IP13'-Tab1, MGC57664, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TGF-beta-activated kinase 1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 405-504 of human TAB1 (Q15750).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10454
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.