Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAB1 Rabbit anti-Human, Mouse, Rat, Clone: 2W0P2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $515.50
Specifications
Antigen | TAB1 |
---|---|
Clone | 2W0P2 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TAB1 Monoclonal antibody specifically detects TAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TAB1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10454 | |
IgG | |
Affinity purified |
2W0P2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
MAP3K7IP13'-Tab1, MGC57664, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TGF-beta-activated kinase 1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 405-504 of human TAB1 (Q15750). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title