Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAB2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB164327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164327 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164327 Supplier Novus Biologicals Supplier No. NBP281735AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAB2 Polyclonal antibody specifically detects TAB2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TAB2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias FLJ21885, KIAA0733CHTD2, MAP3K7IP2mitogen-activated protein kinase kinase kinase 7 interacting protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAB-2, TAK1-binding protein 2, TGF-beta activated kinase 1/MAP3K7 binding protein 2, TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, TGF-beta-activated kinase 1-binding protein 2
Host Species Rabbit
Immunogen TAB2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human TAB2. The immunogen is located within the last 50 amino acids of TAB2. Amino Acid Squence: KTPKTQDTEDDEGA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23118
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.