Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAB2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291538 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291538 Supplier Novus Biologicals Supplier No. NBP146199F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAB2 Polyclonal antibody specifically detects TAB2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen TAB2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias FLJ21885, KIAA0733CHTD2, MAP3K7IP2mitogen-activated protein kinase kinase kinase 7 interacting protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAB-2, TAK1-binding protein 2, TGF-beta activated kinase 1/MAP3K7 binding protein 2, TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, TGF-beta-activated kinase 1-binding protein 2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human TAK1-Binding Protein 2 using the numbering given in entry NP_055908.1 (GeneID 23118).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23118
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.