Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACC3 Antibody (OTI6F1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N274436APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274436APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274436APC7 Supplier Novus Biologicals Supplier No. NBP274436APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TACC3 Monoclonal specifically detects TACC3 in Human, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen TACC3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI6F1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3
Gene Symbols TACC3
Host Species Mouse
Immunogen Full length recombinant protein of human TACC3 (NP_006333) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10460.0
Target Species Human, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.