Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACE/ADAM17 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB018749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018749 Supplier Novus Biologicals Supplier No. NBP177044V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TACE/ADAM17
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TACE. The immunogen is located within the last 50 amino acids of TACE. Amino Acid Squence: ASFKLQRQNRVDSKETE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Myeloid derived Suppressor Cell, Neurodegeneration, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6868
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.