Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACE/ADAM17 Antibody (MM0561-8C13), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP212018Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212018Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212018Y Supplier Novus Biologicals Supplier No. NBP212018AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TACE/ADAM17
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0561-8C13
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Gene Accession No. P78536
Gene Alias ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme
Gene Symbols ADAM17
Host Species Mouse
Immunogen Human recombinant TACE/ADAM17
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6868
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.