Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Antibody (006), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB069070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069070 Supplier Novus Biologicals Supplier No. NBP290025IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for ELISA.

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications ELISA
Classification Monoclonal
Clone 6
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TACI/TNFRSF13B/CVID (O14836-2; Ser2-Thr120).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.