Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Antibody (165604), FITC, Novus Biologicals™
SDP

Catalog No. NB297021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297021 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297021 Supplier Novus Biologicals Supplier No. FAB1741F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TACI/TNFRSF13B/CVID Monoclonal antibody specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOF

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications Flow Cytometry, Blocking Assay, CyTOF
Classification Monoclonal
Clone 165604
Conjugate FITC
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation PBS
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TACI/TNFRSF13B, Ser2-Thr166, Accession # O14836
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.