Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Antibody, Novus Biologicals™
SDP

Catalog No. NB10056516T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056516T 0.025 mg
NB10056516 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056516T Supplier Novus Biologicals Supplier No. NB10056516SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

TACI/TNFRSF13B/CVID Polyclonal specifically detects TACI/TNFRSF13B/CVID in Human, Mouse samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin reported in scientific literature (PMID 22512719)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_036584
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 116-132 of human TACI. Human and mouse TACI amino acid sequences are 65% conserved in the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.