Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB196420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB196420 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB196420 Supplier Novus Biologicals Supplier No. NBP176779AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TACI/TNFRSF13B/CVID Polyclonal antibody specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide mapping at the amino terminus of human TACI. The immunogen is located within the first 50 amino acids of TACI. Amino Acid Squence: SGLGRSRRGGRSRVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.