Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Antibody (MM0562-9D22) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211937 0.1 mg
NB018965 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211937 Supplier Novus Biologicals Supplier No. NBP211937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
Classification Monoclonal
Clone MM0562-9D22
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:20-1:200, Block/Neutralize
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. O14836
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Mouse
Immunogen Human recombinant TACI/TNFRSF13B/CVID (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23495
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.