Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TACI/TNFRSF13B/CVID Rabbit anti-Human, Clone: 6, Novus Biologicals™
SDP

Catalog No. NB021513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021513 100 μg
NB021514 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021513 Supplier Novus Biologicals Supplier No. NBP290025100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for ELISA.

Specifications

Antigen TACI/TNFRSF13B/CVID
Applications ELISA
Classification Monoclonal
Clone 6
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B
Host Species Rabbit
Immunogen Recombinant Human TACI/TNFRSF13B/CVID Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.