Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tacrolimus, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. J63571MCPM
Click to view available options
Quantity:
50 mg
100 mg
500 mg

Tacrolimus also known as FK-506 or fujimycin, is a macrolide compound that exhibits immunosuppressant, neuroprotective, and neuroregenerative activities and can inhibit T-cell proliferation and activation.

General Description

  • Tacrolimus is a macrolide isolated from Streptomyces tsukubaensis, that shows immunosuppressant, neuroprotective, and neuroregenerative activities, having the ability to block T-cell proliferation and activation in response to antigenic or mitogenic stimulation
  • Tacrolimus interacts with the immunophilins cyclophilin and FKBP-12. Tacrolimus binds to the FKBP-12 protein (FK506 binding protein) and forms a complex with calcium-dependent proteins. This FKBP12-FK506 complex inhibits calcineurin phosphatase activity, resulting in decreased cytokine production, inhibition of T-lymphocyte signal transduction
  • Tacrolimus also inhibits the transcription for genes that encode IL-3, IL-4, IL-5, GM-CSF, and TNF, all of which are involved in the early stages of T-cell activation
  • This compound can also inhibit the release of pre-formed mediators from skin mast cells and basophils and downregulate the expression of FceRI on Langerhans cells

Application

  • Tacrolimus has been studied in vitro to understand its impact on neutrophil production and/or survival
  • It is applied to suppress the inflammation in ulcerative cells
  • It is used as a research tool in chemically induced dimerization applications in biology
  • It has been used in in vitro studies to investigate its effect on human monocyte-derived dentritic cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104987-11-3
Molecular Formula C44H69NO12
Synonym FK-506; Fugimycin

Specifications

Quantity 100 mg
Content And Storage -30°C to -10°C
For Use With (Application) Interacts with the immunophilins cyclophilin and FKBP-12
Melting Point 120°C to 125°C
Solubility Freely soluble in DMSO or ethanol; Poorly soluble in water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.