Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TADA1L Antibody (PCRP-TADA1-1C9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB157413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157413 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157413 Supplier Novus Biologicals Supplier No. NBP314264AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TADA1L Monoclonal antibody specifically detects TADA1L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen TADA1L
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-TADA1-1C9
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ADA1, hADA1, HFI1, SPT3-associated factor 42, SPT3-associated factor 42 (STAF42), STAF42transcriptional adaptor 1 (HFI1 homolog, yeast)-like, TADA1LRP1-9E21.4, transcriptional adapter 1, Transcriptional adapter 1-like protein, transcriptional adaptor 1, transcriptional adaptor 1 (HFI1 homolog, yeast)
Host Species Mouse
Immunogen Recombinant full-length human TADA1Lprotein (Uniprot: Q96BN2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117143
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.