Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF15 Antibody (4D71), FITC, Novus Biologicals™
SDP

Catalog No. NBP192685F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192685F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192685F Supplier Novus Biologicals Supplier No. NBP192685F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAF15 Monoclonal specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TAF15
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4D71
Concentration 1mg/mL
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q92804
Gene Alias 68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor
Gene Symbols TAF15
Host Species Mouse
Immunogen Full length recombinant human TAF15 expressed in and purified from E. coli. [UniProt# Q92804]
Molecular Weight of Antigen 68 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8148
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.